siRNA Details
| Gene Symbol: RGD1305110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100971.1 | NP_001094441.1 | 14 | 1705 | ||
| XM_006251544.2 | XP_006251606.1 | 14 | 1704 | |||
| GenBank mRNA | BC128776.1 | - | 14 | 1705 | 7133856 | |
| FQ212361.1 | - | 7 | 780 | |||
| FQ212938.1 | - | 2 | 61 | |||
| FQ213138.1 | - | 2 | 61 | |||
| FQ216884.1 | - | 3 | 290 | |||

RGD1305110
Genomic Map