siRNA Details
| Gene Symbol: RGD1305110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100971.1 | NP_001094441.1 | 11 | 1273 | ||
| XM_006251544.2 | XP_006251606.1 | 11 | 1272 | |||
| GenBank mRNA | BC128776.1 | - | 11 | 1273 | 7133856 | |
| FQ212361.1 | - | 4 | 348 | |||

RGD1305110
Genomic Map