siRNA Details
Gene Symbol: Pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107242.2 | NP_001100712.1 | 2 | 637 | ||
GenBank mRNA | FM088835.1 | - | 1 | 286 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107242.2 | NP_001100712.1 | 2 | 637 | ||
GenBank mRNA | FM088835.1 | - | 1 | 286 |