siRNA Details
| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251597.2 | XP_006251659.1 | 1 | 608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fancl| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251597.2 | XP_006251659.1 | 1 | 608 | ||