siRNA Details
| Gene Symbol: Efemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012039.1 | NP_001012039.1 | 11 | 1348 | ||
| XM_008770456.1 | XP_008768678.1 | 11 | 1448 | |||
| XM_008770457.1 | XP_008768679.1 | 10 | 1368 | |||
| GenBank mRNA | BC079455.1 | - | 11 | 1348 | 7121509 | |
| D89730.1 | - | 9 | 1244 | |||

Efemp1
Genomic Map