siRNA Details
| Gene Symbol: Mtif2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004254.1 | NP_001004254.1 | 14 | 2146 | ||
| GenBank mRNA | BC079421.1 | - | 14 | 2146 | 7113831 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtif2| Gene Symbol: Mtif2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004254.1 | NP_001004254.1 | 14 | 2146 | ||
| GenBank mRNA | BC079421.1 | - | 14 | 2146 | 7113831 | |