siRNA Details
| Gene Symbol: Synpo2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770581.1 | XP_008768803.1 | 2 | 1683 | ||
| XM_223791.8 | XP_223791.6 | 4 | 2041 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synpo2l| Gene Symbol: Synpo2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770581.1 | XP_008768803.1 | 2 | 1683 | ||
| XM_223791.8 | XP_223791.6 | 4 | 2041 | |||