siRNA Details
| Gene Symbol: Samd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012040.1 | NP_001012040.1 | 2 | 731 | ||
| XM_006251640.1 | XP_006251702.1 | 2 | 769 | |||
| GenBank mRNA | BC082100.1 | - | 2 | 731 | 7134467 | |
| FQ219794.1 | - | 2 | 769 | |||

Samd8
Genomic Map