siRNA Details
| Gene Symbol: Lrrc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728176.4 | XP_002728222.1 | 2 | 1299 | ||
| XM_006251719.2 | XP_006251781.1 | 2 | 2803 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc3b| Gene Symbol: Lrrc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728176.4 | XP_002728222.1 | 2 | 1299 | ||
| XM_006251719.2 | XP_006251781.1 | 2 | 2803 | |||