siRNA Details
| Gene Symbol: Nek10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251721.2 | XP_006251783.1 | 13 | 3423 | ||
| XM_008770599.1 | XP_008768821.1 | 9 | 3194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek10| Gene Symbol: Nek10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251721.2 | XP_006251783.1 | 13 | 3423 | ||
| XM_008770599.1 | XP_008768821.1 | 9 | 3194 | |||