siRNA Details
| Gene Symbol: Ddhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033066.1 | NP_001028238.1 | 1 | 814 | ||
| XM_006251769.2 | XP_006251831.1 | 1 | 882 | |||
| XM_006251770.2 | XP_006251832.1 | 1 | 882 | |||
| XM_006251771.2 | XP_006251833.1 | 1 | 882 | |||
| XM_006251772.2 | XP_006251834.1 | 1 | 882 | |||
| XM_006251773.2 | XP_006251835.1 | 1 | 882 | |||
| XM_006251774.2 | XP_006251836.1 | 1 | 881 | |||
| GenBank mRNA | BC103649.1 | - | 1 | 814 | 7314879 | |

Ddhd1
Genomic Map