siRNA Details
Gene Symbol: Atg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107258.1 | NP_001100728.1 | 6, 7 | 911 | ||
GenBank mRNA | FQ223033.1 | - | 6, 7 | 879 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107258.1 | NP_001100728.1 | 6, 7 | 911 | ||
GenBank mRNA | FQ223033.1 | - | 6, 7 | 879 |