siRNA Details
| Gene Symbol: Dcaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009686.2 | NP_001009686.1 | 8 | 1024 | ||
| XM_006251971.2 | XP_006252033.1 | 9 | 1359 | |||
| GenBank mRNA | BC087110.1 | - | 7 | 805 | 7193336 | |
| BC091364.1 | - | 8 | 1024 | 7319156 | ||
| FQ222851.1 | - | 9 | 1325 | |||

Dcaf11
Genomic Map