siRNA Details
| Gene Symbol: Nfatc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107264.1 | NP_001100734.1 | 2 | 573 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfatc4| Gene Symbol: Nfatc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107264.1 | NP_001100734.1 | 2 | 573 | ||