siRNA Details
| Gene Symbol: LOC100912447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770716.1 | XP_008768938.1 | 8 | 4351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100912447| Gene Symbol: LOC100912447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770716.1 | XP_008768938.1 | 8 | 4351 | ||