siRNA Details
| Gene Symbol: Ift88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107266.1 | NP_001100736.1 | 11 | 814 | ||
| XM_006252066.2 | XP_006252128.1 | 11 | 1796 | |||
| XM_006252067.2 | XP_006252129.1 | 12 | 1988 | |||
| XM_006252068.2 | XP_006252130.1 | 11 | 1097 | |||
| XM_006252070.2 | XP_006252132.1 | 11 | 1795 | |||
| XM_006252071.2 | XP_006252133.1 | 11 | 1796 | |||

Ift88
Genomic Map