siRNA Details
| Gene Symbol: Mtmr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107268.1 | NP_001100738.1 | 2 | 216 | ||
| XM_006252111.1 | XP_006252173.1 | 2 | 250 | |||
| XM_006252112.1 | XP_006252174.1 | 2 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr6| Gene Symbol: Mtmr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107268.1 | NP_001100738.1 | 2 | 216 | ||
| XM_006252111.1 | XP_006252173.1 | 2 | 250 | |||
| XM_006252112.1 | XP_006252174.1 | 2 | 250 | |||