siRNA Details
| Gene Symbol: Pinx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083337.1 | NP_001076806.1 | 2 | 90 | ||
| XM_006252181.2 | XP_006252243.1 | 2 | 993 | |||
| GenBank mRNA | BC158553.1 | - | 2 | 189 | 9088212 | |
| EF446165.1 | - | 2 | 90 | |||

Pinx1
Genomic Map