siRNA Details
| Gene Symbol: Fbxo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013132.2 | NP_001013150.1 | 5 | 554 | ||
| XM_006252201.2 | XP_006252263.1 | 6 | 1327 | |||
| XM_006252202.2 | XP_006252264.1 | 5 | 1207 | |||
| GenBank mRNA | BC062801.1 | - | 5 | 557 | 6889991 | |
| FQ213407.1 | - | 5 | 488 | |||

Fbxo16
Genomic Map