siRNA Details
| Gene Symbol: Pnma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107272.1 | NP_001100742.1 | 3 | 1429 | ||
| XM_006252116.2 | XP_006252178.1 | 4 | 1603 | |||
| XM_008770756.1 | XP_008768978.1 | 3 | 1454 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnma2| Gene Symbol: Pnma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107272.1 | NP_001100742.1 | 3 | 1429 | ||
| XM_006252116.2 | XP_006252178.1 | 4 | 1603 | |||
| XM_008770756.1 | XP_008768978.1 | 3 | 1454 | |||