siRNA Details
| Gene Symbol: Cdca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107273.1 | NP_001100743.1 | 15 | 2525 | ||
| XM_006252117.2 | XP_006252179.1 | 15 | 2602 | |||
| XM_006252118.2 | XP_006252180.1 | 15 | 2710 | |||
| XM_006252120.2 | XP_006252182.1 | 15 | 2599 | |||
| XM_006252121.2 | XP_006252183.1 | 12 | 2656 | |||
| XM_008770757.1 | XP_008768979.1 | 12 | 1982 | |||
| GenBank mRNA | FQ211026.1 | - | 15 | 2509 | ||

Cdca2
Genomic Map