siRNA Details
Gene Symbol: Slc25a37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013996.1 | NP_001014018.1 | 4 | 1139 | ||
XM_006252266.1 | XP_006252328.1 | 5 | 1311 | |||
GenBank mRNA | BC082071.1 | - | 4 | 1139 | 7132913 | |
FQ227341.1 | - | 4 | 1083 | |||
FQ227813.1 | - | 1 | 474 | |||
FQ230701.1 | - | 3 | 796 | |||
FQ231945.1 | - | 3 | 627 | |||
FQ233331.1 | - | 4 | 1040 |
