siRNA Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170472.1 | NP_001163943.1 | 10 | 1065 | ||
| XM_006252268.2 | XP_006252330.1 | 10 | 1182 | |||
| GenBank mRNA | CA510912.1 | - | 5 | 457 | ||
| FQ211644.1 | - | 10 | 1176 | |||

Ccar2
Genomic Map