siRNA Details
| Gene Symbol: Pcdh20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107280.1 | NP_001100750.1 | 2 | 308 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdh20| Gene Symbol: Pcdh20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107280.1 | NP_001100750.1 | 2 | 308 | ||