siRNA Details
| Gene Symbol: Klf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107281.1 | NP_001100751.1 | 4 | 684 | ||
| XM_006252408.2 | XP_006252470.1 | 4 | 686 | |||
| XM_006252409.2 | XP_006252471.1 | 4 | 681 | |||
| XM_006252410.2 | XP_006252472.1 | 4 | 752 | |||
| XM_006252413.2 | XP_006252475.1 | 3 | 534 | |||
| XM_006252415.2 | XP_006252477.1 | 3 | 531 | |||
| XM_008770870.1 | XP_008769092.1 | 3 | 2488 | |||
| XM_008770871.1 | XP_008769093.1 | 5 | 698 | |||

Klf12
Genomic Map