siRNA Details
| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107286.1 | NP_001100756.1 | 8 | 931 | ||
| GenBank mRNA | BC166991.1 | - | 5 | 375 | ||
| FQ213394.1 | - | 4 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgds| Gene Symbol: Tgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107286.1 | NP_001100756.1 | 8 | 931 | ||
| GenBank mRNA | BC166991.1 | - | 5 | 375 | ||
| FQ213394.1 | - | 4 | 271 | |||