siRNA Details
| Gene Symbol: Gpr180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006994.1 | NP_001006995.1 | 4 | 617 | ||
| XM_006252452.2 | XP_006252514.1 | 4 | 713 | |||
| GenBank mRNA | BC083689.1 | - | 4 | 617 | 7189368 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr180| Gene Symbol: Gpr180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006994.1 | NP_001006995.1 | 4 | 617 | ||
| XM_006252452.2 | XP_006252514.1 | 4 | 713 | |||
| GenBank mRNA | BC083689.1 | - | 4 | 617 | 7189368 | |