siRNA Details
| Gene Symbol: Duxbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252678.2 | XP_006252740.1 | 3 | 3120 | ||
| XM_006252679.2 | XP_006252741.1 | 4 | 1121 | |||
| XM_008771030.1 | XP_008769252.1 | 1 | 612 | |||
| XM_008771031.1 | XP_008769253.1 | 2 | 3795 | |||
| XM_224643.6 | XP_224643.1 | 3 | 1796 | |||

Duxbl1
Genomic Map