siRNA Details
| Gene Symbol: Dennd6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134467.1 | NP_001127939.1 | 18 | 1618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dennd6a| Gene Symbol: Dennd6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134467.1 | NP_001127939.1 | 18 | 1618 | ||