siRNA Details
| Gene Symbol: Cacna2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175595.2 | NP_783185.1 | 6 | 557 | ||
| GenBank mRNA | AF486278.1 | - | 6 | 557 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna2d3| Gene Symbol: Cacna2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175595.2 | NP_783185.1 | 6 | 557 | ||
| GenBank mRNA | AF486278.1 | - | 6 | 557 | ||