siRNA Details
| Gene Symbol: Cacna2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175595.2 | NP_783185.1 | 20, 21 | 1893 | ||
| XM_008770993.1 | XP_008769215.1 | 11, 12 | 1535 | |||
| XM_008770994.1 | XP_008769216.1 | 9, 10 | 727 | |||
| GenBank mRNA | AF486278.1 | - | 20, 21 | 1893 | ||

Cacna2d3
Genomic Map