siRNA Details
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107290.1 | NP_001100760.1 | 1 | 116 | ||
XM_006252603.2 | XP_006252665.1 | 1 | 102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107290.1 | NP_001100760.1 | 1 | 116 | ||
XM_006252603.2 | XP_006252665.1 | 1 | 102 |