siRNA Details
| Gene Symbol: Itih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107291.2 | NP_001100761.2 | 22 | 2670 | ||
| XM_008770995.1 | XP_008769217.1 | 21 | 2601 | |||
| GenBank mRNA | BC166831.1 | - | 22 | 2670 | 8374222 | |
| FQ219596.1 | - | 6 | 752 | |||

Itih1
Genomic Map