siRNA Details
| Gene Symbol: Itih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107291.2 | NP_001100761.2 | 19 | 2166 | ||
| XM_008770995.1 | XP_008769217.1 | 18 | 2097 | |||
| GenBank mRNA | BC166831.1 | - | 19 | 2166 | 8374222 | |
| FQ219596.1 | - | 3 | 248 | |||

Itih1
Genomic Map