siRNA Details
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013134.2 | NP_001013152.2 | 3 | 522 | ||
| XM_008770996.1 | XP_008769218.1 | 3 | 626 | |||
| GenBank mRNA | BC088323.1 | - | 3 | 534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek4| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013134.2 | NP_001013152.2 | 3 | 522 | ||
| XM_008770996.1 | XP_008769218.1 | 3 | 626 | |||
| GenBank mRNA | BC088323.1 | - | 3 | 534 | ||