siRNA Details
| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107292.1 | NP_001100762.1 | 13 | 1779 | ||
| XM_006252606.2 | XP_006252668.1 | 13 | 1458 | |||
| XM_006252607.2 | XP_006252669.1 | 13 | 1458 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bap1| Gene Symbol: Bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107292.1 | NP_001100762.1 | 13 | 1779 | ||
| XM_006252606.2 | XP_006252668.1 | 13 | 1458 | |||
| XM_006252607.2 | XP_006252669.1 | 13 | 1458 | |||