siRNA Details
| Gene Symbol: Capn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030037.1 | NP_001025208.1 | 6 | 844 | ||
| XM_006252608.2 | XP_006252670.2 | 7 | 1077 | |||
| XM_008770997.1 | XP_008769219.1 | 6 | 1648 | |||
| GenBank mRNA | BC099770.1 | - | 6 | 844 | 7932056 | |

Capn7
Genomic Map