siRNA Details
| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771060.1 | XP_008769282.1 | 1 | 1415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt15| Gene Symbol: Galnt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771060.1 | XP_008769282.1 | 1 | 1415 | ||