siRNA Details
| Gene Symbol: Ercc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107296.1 | NP_001100766.1 | 11 | 2019 | ||
| XM_006252734.2 | XP_006252796.1 | 12 | 2405 | |||
| XM_006252735.2 | XP_006252797.1 | 12 | 2347 | |||
| XM_006252736.2 | XP_006252798.1 | 12 | 2554 | |||
| XM_006252737.2 | XP_006252799.1 | 11 | 2246 | |||
| XM_006252738.1 | XP_006252800.1 | 9 | 1712 | |||

Ercc6
Genomic Map