siRNA Details
| Gene Symbol: Lrrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013999.1 | NP_001014021.1 | 3 | 417 | ||
| XM_008771097.1 | XP_008769319.1 | 3 | 793 | |||
| GenBank mRNA | BC081908.1 | - | 3 | 417 | 7111390 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc18| Gene Symbol: Lrrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013999.1 | NP_001014021.1 | 3 | 417 | ||
| XM_008771097.1 | XP_008769319.1 | 3 | 793 | |||
| GenBank mRNA | BC081908.1 | - | 3 | 417 | 7111390 | |