siRNA Details
| Gene Symbol: Ptpn20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080149.1 | NP_001073618.1 | 8 | 972 | ||
| XM_006252765.2 | XP_006252827.1 | 9 | 1045 | |||
| XM_006252767.2 | XP_006252829.1 | 9 | 1012 | |||
| XM_008771099.1 | XP_008769321.1 | 9 | 998 | |||
| XM_008771100.1 | XP_008769322.1 | 9 | 1045 | |||
| GenBank mRNA | BC129117.1 | - | 8 | 972 | 7114504 | |

Ptpn20b
Genomic Map