siRNA Details
| Gene Symbol: Syt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181632.2 | NP_853663.2 | 5 | 901 | ||
| GenBank mRNA | AB109021.1 | - | 4 | 796 | ||
| BC084685.1 | - | 5 | 901 | 7100793 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt15| Gene Symbol: Syt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181632.2 | NP_853663.2 | 5 | 901 | ||
| GenBank mRNA | AB109021.1 | - | 4 | 796 | ||
| BC084685.1 | - | 5 | 901 | 7100793 | ||