siRNA Details
| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100175.1 | NP_001093645.1 | 3 | 554 | ||
| GenBank mRNA | EF690436.1 | - | 3 | 554 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem38a| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100175.1 | NP_001093645.1 | 3 | 554 | ||
| GenBank mRNA | EF690436.1 | - | 3 | 554 | ||