siRNA Details
| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107310.1 | NP_001100780.1 | 8 | 907 | ||
| XM_008771117.1 | XP_008769339.1 | 9 | 898 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spock3| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107310.1 | NP_001100780.1 | 8 | 907 | ||
| XM_008771117.1 | XP_008769339.1 | 9 | 898 | |||