siRNA Details
| Gene Symbol: Sh3rf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198764.1 | NP_942059.1 | 3 | 720 | ||
| XM_008771225.1 | XP_008769447.1 | 2 | 837 | |||
| GenBank mRNA | AF515735.1 | - | 3 | 720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3rf1| Gene Symbol: Sh3rf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198764.1 | NP_942059.1 | 3 | 720 | ||
| XM_008771225.1 | XP_008769447.1 | 2 | 837 | |||
| GenBank mRNA | AF515735.1 | - | 3 | 720 | ||