siRNA Details
| Gene Symbol: Sh3rf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198764.1 | NP_942059.1 | 1 | 276 | ||
| GenBank mRNA | AF515735.1 | - | 1 | 276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3rf1| Gene Symbol: Sh3rf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198764.1 | NP_942059.1 | 1 | 276 | ||
| GenBank mRNA | AF515735.1 | - | 1 | 276 | ||