siRNA Details
| Gene Symbol: Mfap3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012049.1 | NP_001012049.1 | 3 | 1356 | ||
| GenBank mRNA | BC078969.1 | - | 3 | 1356 | 7111774 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfap3l| Gene Symbol: Mfap3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012049.1 | NP_001012049.1 | 3 | 1356 | ||
| GenBank mRNA | BC078969.1 | - | 3 | 1356 | 7111774 | |