siRNA Details
| Gene Symbol: Gpm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178105.2 | NP_835206.1 | 4, 5 | 572 | ||
| XM_006253093.2 | XP_006253155.1 | 5, 6 | 1408 | |||
| GenBank mRNA | AB089242.1 | - | 4, 5 | 551 | ||
| BC088862.1 | - | 4, 5 | 572 | 7313006 | ||
| FQ211804.1 | - | 5, 6 | 1027 | |||

Gpm6a
Genomic Map