siRNA Details
| Gene Symbol: Tenm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169133.2 | NP_001162604.1 | 26 | 5597 | ||
| XM_006253115.2 | XP_006253177.1 | 24 | 5581 | |||
| XM_008771244.1 | XP_008769466.1 | 27 | 5650 | |||
| XM_008771245.1 | XP_008769467.1 | 27 | 5647 | |||

Tenm3
Genomic Map