siRNA Details
| Gene Symbol: Stox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134863.1 | NP_001128335.1 | 3 | 3205 | ||
| XM_006253151.2 | XP_006253213.2 | 3 | 2488 | |||
| XM_006253152.2 | XP_006253214.1 | 3 | 1916 | |||
| XM_006253153.2 | XP_006253215.1 | 3 | 2296 | |||
| XM_008771258.1 | XP_008769480.1 | 3 | 2056 | |||

Stox2
Genomic Map